4-Nitro-3-(trifluormethyl)phenol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
α,α,α-Trifluoro-4-nitro-m-cresol, 5-Hydroxy-2-nitrobenzotrifluoride
N0279-5G
N0279-25G
TCIAN0279-5GEA
542
DKK
TCIAN0279-5G
TCIAN0279-25G
4-Nitro-3-(trifluormethyl)phenol ≥99.0% (by GC)
4-Nitro-3-(trifluormethyl)phenol
Formel:
C₇H₄F₃NO₃ Kogepunkt: 135 - 138 °C (0,01 -rr) Smeltepunkt: 81 °C |
MDL Number:
MFCD00014714 CAS-nummer: 88-30-2 EINECS: 201-818-2 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,09253 |
|
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 79.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...