5-Fluor-2-methoxybenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Cyano-4-fluoroanisole

F0479-1G F0479-5G
TCIAF0479-1GEA 181 DKK
TCIAF0479-1G TCIAF0479-5G
5-Fluor-2-methoxybenzonitril ≥98.0% (by GC)
5-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
Molekylvægt: 151.14 g/mol
MDL Number: MFCD02683503
CAS-nummer: 189628-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06b0c9842a950d510ff97c30c98138a7de122ca4&vid=06b0c9842a950d510ff97c30c98138a7de122ca4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 119.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR