3-(Trifluormethyl)phenylacetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Trifluoromethyl)benzyl cyanide

T2367-25G T2367-5G
TCIAT2367-25GEA 673 DKK
TCIAT2367-25G TCIAT2367-5G
3-(Trifluormethyl)phenylacetonitril ≥98.0% (by GC)
3-(Trifluormethyl)phenylacetonitril
Formel: C₉H₆F₃N
Molekylvægt: 185.15 g/mol
Kogepunkt: 93°C
Massefylde: 1.18
Flammepunkt: 48°C
MDL Number: MFCD00001913
CAS-nummer: 2338-76-3
EINECS: 219-053-8
UN nummer: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=046fb2ec7d4a14853d44ab29971c6236d330f056&vid=046fb2ec7d4a14853d44ab29971c6236d330f056&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR