2-Fluor-6-trifluormethylpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
F0826-25G
F0826-5G
TCIAF0826-25GEA
1030
DKK
TCIAF0826-25G
TCIAF0826-5G
2-Fluor-6-trifluormethylpyridin ≥98.0% (by GC)
2-Fluor-6-trifluormethylpyridin
Formel:
C₆H₃F₄N |
MDL Number:
MFCD03092901 CAS-nummer: 94239-04-0 EINECS: 428-100-3 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...