Thiobenzamid ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Benzothioamide

T0194-25G T0194-100G
TCIAT0194-25GEA 460 DKK
TCIAT0194-25G TCIAT0194-100G
Thiobenzamid ≥98.0% (by HPLC, total nitrogen)
Thiobenzamid
Formel: C₇H₇NS
Molekylvægt: 137.21 g/mol
Kogepunkt: 245
MDL Number: MFCD00008060
CAS-nummer: 2227-79-4
EINECS: 218-765-6
UN nummer: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8f9232eaa82a09edb13099a71ad03fb74958a9c&vid=a8f9232eaa82a09edb13099a71ad03fb74958a9c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 115.0 to 119.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR