3,4,5-Trimethoxybenzamid ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T0718-10G
T0718-25G
TCIAT0718-10GEA
288
DKK
TCIAT0718-10G
TCIAT0718-25G
3,4,5-Trimethoxybenzamid ≥98.0% (by HPLC, total nitrogen)
3,4,5-Trimethoxybenzamid
Formel:
C₁₀H₁₃NO₄ |
MDL Number:
MFCD00017129 CAS-nummer: 3086-62-2 EINECS: 221-406-6 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...