3-Nitrobenzamid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
N0321-25G
N0321-500G
TCIAN0321-25GEA
353
DKK
TCIAN0321-25G
TCIAN0321-500G
3-Nitrobenzamid ≥98.0% (by GC)
3-Nitrobenzamid
Formel:
C₇H₆N₂O₃ Molekylvægt: 166.14 g/mol Kogepunkt: 310 Smeltepunkt: 140 |
MDL Number:
MFCD00007984 CAS-nummer: 645-09-0 EINECS: 211-431-0 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 143.0 to 146.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...