Prednison ≥96.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1276-5G P1276-25G
TCIAP1276-5GEA 460 DKK
TCIAP1276-5G TCIAP1276-25G
Prednison ≥96.0% (by HPLC)
Prednison
Formel: C₂₁H₂₆O₅
Smeltepunkt: 230…232 °C
MDL Number: MFCD00003608
CAS-nummer: 53-03-2
EINECS: 200-160-3
Merck Index: 14,07722
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cd6c6e6c820af7d37f8364a695c6b235d0a0644&vid=2cd6c6e6c820af7d37f8364a695c6b235d0a0644&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Specific rotation [a]20/D +165.0 to +175.0 deg (C=0.5, Dioxane)
Solubility in N,N-DMF very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR