Hydrocortison ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H0533-25G H0533-1G
TCIAH0533-25GEA 1970 DKK
TCIAH0533-25G TCIAH0533-1G
Hydrocortison ≥98.0% (by HPLC)
Hydrocortison
Formel: C₂₁H₃₀O₅
Molekylvægt: 362.47 g/mol
Kogepunkt: 567
MDL Number: MFCD00011654
CAS-nummer: 50-23-7
EINECS: 200-020-1
Merck Index: 14,04787

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +150 to +156 deg (C=1, Dioxane)
Solubility in hot Dioxane almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR