4-(Dimethylamino)benzophenon ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2561-25G D2561-5G
TCIAD2561-25GEA 903 DKK
TCIAD2561-25G TCIAD2561-5G
4-(Dimethylamino)benzophenon ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)benzophenon
Formel: C₁₅H₁₅NO
Kogepunkt: 250°C
Smeltepunkt: 97…98 °C
MDL Number: MFCD00008311
CAS-nummer: 530-44-9
EINECS: 208-478-4
Merck Index: 14,03233
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a118165ceb916519b463777a3cf747e5af42b8f&vid=3a118165ceb916519b463777a3cf747e5af42b8f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 89.0 to 93.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR