4-Aminobenzamidin, 2 HCl ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride

A2115-25G A2115-5G
TCIAA2115-25GEA 1420 DKK
TCIAA2115-25G TCIAA2115-5G
4-Aminobenzamidin, 2 HCl ≥98.0% (by total nitrogen basis)
4-Aminobenzamidin, 2 HCl
Formel: C₇H₁₁Cl₂N₃
Smeltepunkt: 291°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00013001
CAS-nummer: 2498-50-2
EINECS: 219-692-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee684b05ca4fb8b10328f255f8ab2e22be1d341a&vid=ee684b05ca4fb8b10328f255f8ab2e22be1d341a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR