Benzamidin, HCl ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

B0013-25G B0013-500G B0013-100G
TCIAB0013-25GEA 493 DKK
TCIAB0013-25G TCIAB0013-500G TCIAB0013-100G
Benzamidin, HCl ≥96.0% (by titrimetric analysis)
Benzamidin, HCl
Formel: C₇H₉ClN₂
Molekylvægt: 156.61 g/mol
MDL Number: MFCD00066285
CAS-nummer: 1670-14-0
EINECS: 216-795-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aabcd643dd9da1ea28122459bcc5dddf190d169&vid=1aabcd643dd9da1ea28122459bcc5dddf190d169&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 96.0 % (after deducting NH4Cl)
Melting point 169.0 to 173.0 deg-C (after drying)
Solubility in Water almost transparency
Water max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR