4-Chlor-3-nitrobenzensulfonylchlorid ≥96.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Chloro-3-nitrobenzenesulphonyl Chloride, 4-Chloro-3-nitrobenzene-1-sulphonyl chloride

C1887-25G C1887-5G
TCIAC1887-25GEA 2080 DKK
TCIAC1887-25G TCIAC1887-5G
4-Chlor-3-nitrobenzensulfonylchlorid ≥96.0% (by GC, titration analysis)
4-Chlor-3-nitrobenzensulfonylchlorid
Formel: C₆H₃Cl₂NO₄S
MDL Number: MFCD00007440
CAS-nummer: 97-08-5
EINECS: 202-558-2
UN nummer: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e269d3f77425653e78604a83a700a44e65efdf4&vid=3e269d3f77425653e78604a83a700a44e65efdf4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR