Methyl2-chloracetoacetat ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C1219-25G
C1219-250G
TCIAC1219-25GEA
476
DKK
TCIAC1219-25G
TCIAC1219-250G
Methyl2-chloracetoacetat ≥95.0% (by GC)
Methyl2-chloracetoacetat
Formel:
C₅H₇ClO₃ Kogepunkt: 86°C Smeltepunkt: –33 °C Massefylde: 1,23 g/cm³ (20 °C) Flammepunkt: 71°C |
MDL Number:
MFCD00008757 CAS-nummer: 4755-81-1 EINECS: 225-286-6 UN nummer: 1760 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...