1-Brom-4-Fluor-3-nitrobenzen ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene
B2075-1G
B2075-25G
B2075-5G
TCIAB2075-1GEA
132
DKK
TCIAB2075-1G
TCIAB2075-25G
TCIAB2075-5G
1-Brom-4-Fluor-3-nitrobenzen ≥96.0% (by GC)
1-Brom-4-Fluor-3-nitrobenzen
Formel:
C₆H₃BrFNO₂ Kogepunkt: 147°C Smeltepunkt: 19 °C Massefylde: 1,786 g/cm³ (20 °C) |
MDL Number:
MFCD00129165 CAS-nummer: 364-73-8 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...