5-Brom-2-Fluorbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B2425-25G B2425-5G
TCIAB2425-25GEA 788 DKK
TCIAB2425-25G TCIAB2425-5G
5-Brom-2-Fluorbenzonitril ≥98.0% (by GC)
5-Brom-2-Fluorbenzonitril
Formel: C₇H₃BrFN
Kogepunkt: 255 °C (1013 hPa)
Smeltepunkt: 75…80 °C
MDL Number: MFCD00143424
CAS-nummer: 179897-89-3
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f7bfe139e5505e39b43af0806e1509ba344cb04&vid=3f7bfe139e5505e39b43af0806e1509ba344cb04&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR