1-Brom-2-Chlor-3-Fluorbenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-3-fluorobromobenzene
B3119-25G
B3119-5G
TCIAB3119-25GEA
1050
DKK
TCIAB3119-25G
TCIAB3119-5G
1-Brom-2-Chlor-3-Fluorbenzen ≥98.0% (by GC)
1-Brom-2-Chlor-3-Fluorbenzen
Formel:
C₆H₃BrClF Molekylvægt: 209.44 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04038306 CAS-nummer: 883499-24-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Freezing point | 27.0 to 31.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...