Acetonitril ≥99.5% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

A0060-25ML A0060-500ML
TCIAA0060-25MLEA 132 DKK
TCIAA0060-25ML TCIAA0060-500ML
Acetonitril ≥99.5% (by GC)
Acetonitril
Formel: H₃CCN
Molekylvægt: 41.05 g/mol
Kogepunkt: 82°C
Smeltepunkt: –46°C
Flammepunkt: 10°C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001878
CAS-nummer: 75-05-8
EINECS: 200-835-2
UN nummer: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 14,00070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=448a84cb7a60315bb069f40c31fefc6339139261&vid=359b9e697488014837bd468b45d24b2e731d3e3f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR