Undecanal ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

U0009-25ML U0009-250ML
TCIAU0009-25MLEA 230 DKK
TCIAU0009-25ML TCIAU0009-250ML
Undecanal ≥97.0% (by GC)
Undecanal
Formel: C₁₁H₂₂O
Kogepunkt: 80°C
Smeltepunkt: –2 °C
Massefylde: 0,827 g/cm³ (20 °C)
Flammepunkt: 113°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007016
CAS-nummer: 112-44-7
EINECS: 203-972-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47877d1396194b4a3eff282fa71d88367a2589cd&vid=b28907e123bd839ee9a26abaeec0c88283a2641d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Specific gravity (20/20) 0.8280 to 0.8380


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR