Heptanal ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H0025-25ML
H0025-500ML
TCIAH0025-25MLEA
132
DKK
TCIAH0025-25ML
TCIAH0025-500ML
Heptanal ≥95.0% (by GC)
Heptanal
Formel:
C₇H₁₄O Kogepunkt: 155°C Smeltepunkt: –43 °C Massefylde: 0,82 g/cm³ (20 °C) Flammepunkt: 34°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007028 CAS-nummer: 111-71-7 EINECS: 203-898-4 UN nummer: 3056 ADR: 3,III Merck Index: 14,04658 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...