Butyraldehyd ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B0751-25ML
B0751-500ML
TCIAB0751-25MLEA
148
DKK
TCIAB0751-25ML
TCIAB0751-500ML
Butyraldehyd ≥98.0% (by GC)
Butyraldehyd
Formel:
C₄H₈O Kogepunkt: 76 °C (1013 hPa) Smeltepunkt: –99 °C Massefylde: 0,803 g/cm³ (20 °C) Flammepunkt: –7°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00007023 CAS-nummer: 123-72-8 EINECS: 204-646-6 UN nummer: 1129 ADR: 3,II Merck Index: 14,01591 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...