3-Brom-1,1,1-trifluoracetone ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B1240-25G
B1240-5G
TCIAB1240-25GEA
854
DKK
TCIAB1240-25G
TCIAB1240-5G
3-Brom-1,1,1-trifluoracetone ≥95.0% (by GC)
3-Brom-1,1,1-trifluoracetone
Formel:
BrH₂CC(O)CF₃ Molekylvægt: 190.95 g/mol Kogepunkt: 88°C Massefylde: 1.8389 Flammepunkt: 5°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00039237 CAS-nummer: 431-35-6 EINECS: 207-071-9 UN nummer: 2924 ADR: 3 II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...