1-Bromtetradecan ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Myristyl bromide
B0233-25ML
B0233-500ML
TCIAB0233-25MLEA
132
DKK
TCIAB0233-25ML
TCIAB0233-500ML
1-Bromtetradecan ≥97.0% (by GC)
1-Bromotetradecane
Formel:
BrH₂C(CH₂)₁₂CH₃ Kogepunkt: 205°C Smeltepunkt: 5 °C Massefylde: 1,02 g/cm³ (20 °C) Flammepunkt: 158°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000228 CAS-nummer: 112-71-0 EINECS: 203-999-3 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...