1-Bromhexan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Hexyl bromide

B0600-25G B0600-500G
TCIAB0600-25GEA 132 DKK
TCIAB0600-25G TCIAB0600-500G
1-Bromhexan ≥98.0% (by GC)
1-Bromohexane
Formel: BrH₂C(CH₂)₄CH₃
Kogepunkt: 155°C
Smeltepunkt: -85°C
Massefylde: 1,167 g/cm³ (20 °C)
Flammepunkt: 52°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000271
CAS-nummer: 111-25-1
EINECS: 203-850-2
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d266fe74b4fa46d549d04b206ebfcbd60bfec55&vid=064b68750d08e4cec0eb258f895485e854f97945&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR