1-Bromdodecan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B0587-25ML B0587-500ML
TCIAB0587-25MLEA 140 DKK
TCIAB0587-25ML TCIAB0587-500ML
1-Bromdodecan ≥98.0% (by GC)
1-Bromododecane
Formel: BrH₂C(CH₂)₁₀CH₃
Kogepunkt: 140°C
Smeltepunkt: –10 °C
Massefylde: 1,04 g/cm³ (20 °C)
Flammepunkt: 146°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000225
CAS-nummer: 143-15-7
EINECS: 205-587-9
Merck Index: 14,05389
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfb59191642c53f1ecfb64b8c3ca6dcbe8d0ea34&vid=2b7f99f77c178275165e3f4e8df70a39b9335352&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR