1-Bromdocosan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Docosyl bromide
B0392-25G
B0392-250G
TCIAB0392-25GEA
534
DKK
TCIAB0392-25G
TCIAB0392-250G
1-Bromdocosan ≥98.0% (by GC)
1-Bromodocosane
Formel:
BrH₂C(CH₂)₂₀CH₃ Molekylvægt: 389.5 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00013543 CAS-nummer: 6938-66-5 EINECS: 230-069-4 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 46.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...