1,4-Dibrombutan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D0176-25G D0176-500G
TCIAD0176-25GEA 132 DKK
TCIAD0176-25G TCIAD0176-500G
1,4-Dibrombutan ≥98.0% (by GC)
1,4-Dibromobutane
Formel: BrH₂C(CH₂)₂CH₂Br
Kogepunkt: 198°C
Smeltepunkt: –20 °C
Massefylde: 1,83 g/cm³ (20 °C)
Flammepunkt: 131°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000261
CAS-nummer: 110-52-1
EINECS: 203-775-5
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dcd881d7fc38d05ac6adaac07f990d62ddaa649&vid=29b4ebb2b2e77d67c20dcc2983543f8b44e0fe11&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR