1,1,2,2-Tetrabromethan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T0034-25G
T0034-500G
TCIAT0034-25GEA
181
DKK
TCIAT0034-25G
TCIAT0034-500G
1,1,2,2-Tetrabromethan ≥98.0% (by GC)
1,1,2,2-Tetrabromoethane
Formel:
Br₂HCCHBr₂ Kogepunkt: 243°C Smeltepunkt: 0°C Massefylde: 2,9656 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000133 CAS-nummer: 79-27-6 EINECS: 201-191-5 UN nummer: 2504 ADR: 6.1,III Merck Index: 14,09185 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...