(R)-(+)-Limonen ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexen, (R)-(+)-Dipenten, (R)-(+)-p-Mentha-1,8-dien, D(+)-Limonen

L0047-25ML L0047-500ML
TCIAL0047-25MLEA 148 DKK
TCIAL0047-25ML TCIAL0047-500ML
(R)-(+)-Limonen ≥95.0% (by GC)
(R)-(+)-Limonen
Formel: C₁₀H₁₆
Molekylvægt: 136.24 g/mol
Kogepunkt: 176°C
Smeltepunkt: –75°C
Flammepunkt: 51°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00062991
CAS-nummer: 5989-27-5
EINECS: 227-813-5
UN nummer: 2319
ADR: 3 III
Merck Index: 14,05493
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=648537748c75fa1844827d6f323d5fd2fc48ed9a&vid=3e19356a66e88505004ebe66d0717121ccd2b3c4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Optical purity (GC) min. 98.0 ee%

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR