L(-)-Fenchon ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: (1R)-(-)-Fenchone
F0164-25ML F0164-500ML
TCIAF0164-25MLEA 140 DKK
TCIAF0164-25ML TCIAF0164-500ML
L(-)-Fenchon ≥98.0% (by GC)
L(-)-Fenchon
Formel: C₁₀H₁₆O
Kogepunkt: 193°C
Massefylde: 0,962 g/cm³ (0 °C)
Flammepunkt: 52°C
MDL Number: MFCD00151104
CAS-nummer: 7787-20-4
EINECS: 232-107-5
UN nummer: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f9d3110b7b26eb6529ef9945d8e073c2c58564a&vid=45d98c533b23b5ceb4c4967dc444f45ea0b3a56f&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 80.0 ee%