D(+)-Kamfer ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (1R,4R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R,4R)-(+)-1,7,7-Trimethyl-2-norbornanone, (R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (+)-Camphor
C0010-25G
C0010-500G
TCIAC0010-25GEA
132
DKK
TCIAC0010-25G
TCIAC0010-500G
D(+)-Kamfer ≥98.0% (by GC)
D(+)-Kamfer
Formel:
C₁₀H₁₆O Kogepunkt: 204°C Smeltepunkt: 174…175 °C Massefylde: 0,992 g/cm³ (20 °C) Flammepunkt: 66 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064149 CAS-nummer: 464-49-3 EINECS: 207-355-2 UN nummer: 2717 ADR: 4.1,III Merck Index: 14,01732 |
|
Specifikationer Testresultater
Appearance | White or Colorless powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 177.0 to 182.0 deg-C |
Specific rotation [a]20/D | +41.0 to +46.0 deg (C=20, EtOH) |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...