Dinatrium α-glycerophosphat hydrat ≥85.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

G0096-25G G0096-5G
TCIAG0096-25GEA 1930 DKK
TCIAG0096-25G TCIAG0096-5G
Dinatrium α-glycerophosphat hydrat ≥85.0% (by titrimetric analysis)
Formel: C₃H₇Na₂O₆P·xH₂O
MDL Number: MFCD00035539
CAS-nummer: 1555-56-2
EINECS: 216-304-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05f75d6bc6e7e94278ff627c7a3d4e728b12ed8f&vid=05f75d6bc6e7e94278ff627c7a3d4e728b12ed8f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Iodometric Titration) min. 85.0 % (calcd.on anh.substance)
Water max. 35.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR