2,7-Diaminofluoren, 2 HCl ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Fluorene-2,7-diyldiamine dihydrochloride, 2,7-Fluorenediammonium dichloride, Fluorene-2,7-diamine dihydrochloride

D0093-25G D0093-5G
TCIAD0093-25GEA 2180 DKK
TCIAD0093-25G TCIAD0093-5G
2,7-Diaminofluoren, 2 HCl ≥98.0% (by HPLC)
2,7-Diaminofluoren, 2 HCl
Formel: C₁₃H₁₄Cl₂N₂
Molekylvægt: 269.17 g/mol
MDL Number: MFCD00001128
CAS-nummer: 13548-69-1
EINECS: 236-924-8
Merck Index: 12,04191
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6363f99155b2c306dfb998ea860467645a2c10ac&vid=6363f99155b2c306dfb998ea860467645a2c10ac&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR