Phthalide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Isobenzofuran-1(3H)-one

P0401-25G P0401-500G
TCIAP0401-25GEA 132 DKK
TCIAP0401-25G TCIAP0401-500G
Phthalide ≥98.0% (by GC)
Phthalide
Formel: C₈H₆O₂
Kogepunkt: 290 °C (1013 hPa)
Smeltepunkt: 73 °C
Massefylde: 1,1 g/cm³ (30 °C)
MDL Number: MFCD00005906
CAS-nummer: 87-41-2
EINECS: 201-744-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe7e5390c218911ebafbb70c5fea325a8d66e58&vid=dfe7e5390c218911ebafbb70c5fea325a8d66e58&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR