2-Fluor-6-(trifluormethyl)benzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Cyano-3-fluorobenzotrifluoride
F0701-1G
F0701-5G
TCIAF0701-1GEA
279
DKK
TCIAF0701-1G
TCIAF0701-5G
2-Fluor-6-(trifluormethyl)benzonitril ≥98.0% (by GC)
2-Fluor-6-(trifluormethyl)benzonitril
Formel:
C₈H₃F₄N Molekylvægt: 189.11 g/mol Kogepunkt: 212 Massefylde: 1.373 |
MDL Number:
MFCD00061153 CAS-nummer: 133116-83-3 UN nummer: 3439 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...