2-Chlor-5-(trifluormethyl)benzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Chloro-5-trifluoromethylbenzonitrile, 4-Chloro-3-cyanobenzotrifluoride
C2050-25G
C2050-5G
TCIAC2050-25GEA
887
DKK
TCIAC2050-25G
TCIAC2050-5G
2-Chlor-5-(trifluormethyl)benzonitril ≥98.0% (by GC)
2-Chlor-5-(trifluormethyl)benzonitril
Formel:
C₈H₃ClF₃N Molekylvægt: 205.57 Kogepunkt: 212°C Massefylde: 1.48 |
MDL Number:
MFCD00019742 CAS-nummer: 328-87-0 EINECS: 206-338-7 UN nummer: 3439 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...