4-Acetylbenzonitril ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Methyl p-cyanophenyl ketone, 4'-Cyanoacetophenone, 4-Acetylbenzenecarbonitrile
C1141-10G
C1141-25G
TCIAC1141-10GEA
468
DKK
TCIAC1141-10G
TCIAC1141-25G
4-Acetylbenzonitril ≥97.0% (by GC)
4-Acetylbenzonitril
Formel:
C₉H₇NO Kogepunkt: 96°C Smeltepunkt: 57…58 °C |
MDL Number:
MFCD00001825 CAS-nummer: 1443-80-7 EINECS: 215-885-0 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 56.0 to 59.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...