Hexaammincobalttrichlorid ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

H0051-25G H0051-500G
TCIAH0051-25GEA 304 DKK
TCIAH0051-25G TCIAH0051-500G
Hexaammincobalttrichlorid ≥99.0% (by titrimetric analysis)
Hexaammincobalttrichlorid
Formel: H₁₈CoCl₃N₆
Molekylvægt: 267.47 g/mol
MDL Number: MFCD00036304
CAS-nummer: 10534-89-1
EINECS: 234-103-9
Merck Index: 14,04675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51568fb4a8143fee338f6daec386adc6959edb35&vid=51568fb4a8143fee338f6daec386adc6959edb35&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR