Methyl-2-chloro-4-nitrobenzoate ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-4-nitrobenzoic acid methyl ester, Methyl 2-chloro-4-nitrobenzoate
M1724-25G
M1724-5G
TCIAM1724-25GEA
1070
DKK
TCIAM1724-25G
TCIAM1724-5G
Methyl-2-chloro-4-nitrobenzoate ≥99.0% (by GC)
Methyl-2-chloro-4-nitrobenzoate
Formel:
C₈H₆ClNO₄ |
MDL Number:
MFCD00017013 CAS-nummer: 13324-11-3 EINECS: 236-362-3 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...