2-Thiophencarbonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Thienyl cyanide, Thiophene-2-carbonitrile, 2-Cyanothiophene
T2121-25G
T2121-5G
TCIAT2121-25GEA
517
DKK
TCIAT2121-25G
TCIAT2121-5G
2-Thiophencarbonitril ≥98.0% (by GC)
2-Thiophencarbonitril
Formel:
C₅H₃NS Kogepunkt: 192°C Massefylde: 1,18 g/cm³ (15 °C) Flammepunkt: 53°C |
MDL Number:
MFCD00005416 CAS-nummer: 1003-31-2 EINECS: 213-706-0 UN nummer: 1992 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...