4-Methyl-2,3,5,6-tetrabromphenol ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,3,5,6-Tetrabromo-4-methylphenol, 2,3,5,6-Tetrabromo-p-cresol, 4-Hydroxy-2,3,5,6-tetrabromotoluene
T1430-25G
T1430-5G
TCIAT1430-25GEA
1310
DKK
TCIAT1430-25G
TCIAT1430-5G
4-Methyl-2,3,5,6-tetrabromphenol ≥97.0% (by GC)
4-Methyl-2,3,5,6-tetrabromphenol
Formel:
C₇H₄Br₄O Kogepunkt: 356 °C (1013 hPa) Smeltepunkt: 209 °C |
MDL Number:
MFCD00060318 CAS-nummer: 37721-75-8 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 198.0 to 202.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...