3-Nitro-p-toluonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Methyl-3-nitrobenzonitrile
M1946-25G
M1946-5G
TCIAM1946-25GEA
1370
DKK
TCIAM1946-25G
TCIAM1946-5G
3-Nitro-p-toluonitril ≥98.0% (by GC)
3-Nitro-p-toluonitril
Formel:
C₈H₆N₂O₂ Molekylvægt: 162.15 Kogepunkt: 171°C Smeltepunkt: 107 |
MDL Number:
MFCD00031482 CAS-nummer: 939-79-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...