1-Butylpyridiniumchlorid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Butylpyridinium Chloride, N-Butylpyridinium chloride
B1329-10G
B1329-25G
TCIAB1329-10GEA
386
DKK
TCIAB1329-10G
TCIAB1329-25G
1-Butylpyridiniumchlorid ≥98.0% (by titrimetric analysis)
1-Butylpyridinium chloride
Formel:
C₉H₁₄ClN Molekylvægt: 171.67 g/mol |
MDL Number:
MFCD00060047 CAS-nummer: 1124-64-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...