4-Amino-3-Fluorbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A1965-25G A1965-5G
TCIAA1965-25GEA 3720 DKK
TCIAA1965-25G TCIAA1965-5G
4-Amino-3-Fluorbenzonitril ≥98.0% (by GC)
4-Amino-3-Fluorbenzonitril
Formel: C₇H₅FN₂
MDL Number: MFCD00055559
CAS-nummer: 63069-50-1
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde1370e95a6ac836fec788d7d2d9ff0c0649ac4&vid=bde1370e95a6ac836fec788d7d2d9ff0c0649ac4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR