(4-Nitrophenyl)acetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Nitrobenzyl cyanide

N0229-500G N0229-25G
TCIAN0229-500GEA 4000 DKK
TCIAN0229-500G TCIAN0229-25G
(4-Nitrophenyl)acetonitril ≥98.0% (by GC)
(4-Nitrophenyl)acetonitril
Formel: C₈H₆N₂O₂
Molekylvægt: 162.15 g/mol
Kogepunkt: 197°C
MDL Number: MFCD00007372
CAS-nummer: 555-21-5
EINECS: 209-085-0
UN nummer: 3439
ADR: 6.1 III
Merck Index: 14,06590
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba9a319f45ba20c3f48344a08c62b70be1053b8a&vid=ba9a319f45ba20c3f48344a08c62b70be1053b8a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 118.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR