2-Fluor-4-hydroxybenzonitril ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
F0462-25G
F0462-5G
TCIAF0462-25GEA
1040
DKK
TCIAF0462-25G
TCIAF0462-5G
2-Fluor-4-hydroxybenzonitril ≥98.0% (by GC, titration analysis)
2-Fluor-4-hydroxybenzonitril
Formel:
C₇H₄FNO Kogepunkt: 261 °C (1013 hPa) Smeltepunkt: ∼124 °C Massefylde: 1,45 g/cm³ (20 °C) |
MDL Number:
MFCD00192177 CAS-nummer: 82380-18-5 EINECS: 422-810-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...