2-Amino-6-Fluorbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Cyano-3-fluoroaniline

A2344-25G A2344-5G
TCIAA2344-25GEA 1020 DKK
TCIAA2344-25G TCIAA2344-5G
2-Amino-6-Fluorbenzonitril ≥98.0% (by GC)
2-Amino-6-Fluorbenzonitril
Formel: C₇H₅FN₂
Kogepunkt: 272 °C (1013 hPa)
Smeltepunkt: 127 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00015477
CAS-nummer: 77326-36-4
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=612854b4fac3acf662a0d39a7ac2388962a02c3d&vid=612854b4fac3acf662a0d39a7ac2388962a02c3d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR