Phenylacetylchlorid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P0129-25G P0129-500G P0129-100G
TCIAP0129-25GEA 181 DKK
TCIAP0129-25G TCIAP0129-500G TCIAP0129-100G
Phenylacetylchlorid ≥98.0% (by GC, titration analysis)
Phenylacetylchlorid
Formel: C₈H₇ClO
Molekylvægt: 154.6 g/mol
Kogepunkt: 211°C
Smeltepunkt: 91
Flammepunkt: 102°C
MDL Number: MFCD00000729
CAS-nummer: 103-80-0
EINECS: 203-146-5
UN nummer: 2577
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9d3296260517e0e331be30bcf71b4fbc6918a71&vid=683b46a2daab13b64171cabfd5fc762058ebf1b3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR