Pentyl hexanoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Caproic acid amyl ester, Amyl caproate, Hexanoic acid amyl ester, Caproic acid pentyl ester, Hexanoic acid pentyl ester, Amyl hexanoate, Pentyl hexanoate, Pentyl caproate

H0106-25ML H0106-500ML
TCIAH0106-25MLEA 156 DKK
TCIAH0106-25ML TCIAH0106-500ML
Pentyl hexanoat ≥98.0% (by GC)
Pentyl hexanoat
Formel: C₁₁H₂₂O₂
Kogepunkt: 225 °C (1013 hPa)
Smeltepunkt: –47 °C
Massefylde: 0,858 g/cm³ (25 °C)
Flammepunkt: 91°C
MDL Number: MFCD00027281
CAS-nummer: 540-07-8
EINECS: 208-732-4
Merck Index: 14,00605
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc8b483373c2cb5e83369a65defd3325d860d7e6&vid=fc8b483373c2cb5e83369a65defd3325d860d7e6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR