Dichloracetylchlorid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D0313-25G D0313-500G
TCIAD0313-25GEA 255 DKK
TCIAD0313-25G TCIAD0313-500G
Dichloracetylchlorid ≥98.0% (by GC, titration analysis)
Dichloracetylchlorid
Formel: C₂HCl₃O
Kogepunkt: 108°C
Massefylde: 1,53 g/cm³ (20 °C)
Flammepunkt: 66 °C
MDL Number: MFCD00000840
CAS-nummer: 79-36-7
EINECS: 201-199-9
UN nummer: 1765
ADR: 8,II
Merck Index: 14,03053
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9585b3eeacb823d5a34eaf70f33ba3a29c5a5af&vid=c9585b3eeacb823d5a34eaf70f33ba3a29c5a5af&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR